Drum stands, PE-FB
Lieferant: LACONT UMWELTTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
P70-7114-A P70-7111-A P70-7113-A P70-7112-A P70-7115-A
LACOP70-7114-AEA 0 EUR InStock
LACOP70-7114-A LACOP70-7111-A LACOP70-7113-A LACOP70-7112-A LACOP70-7115-A
Drum stands, PE-FB
Fasslager
PE, black or yellow.
  • 1-drum models made of black PE
  • 2-drum models made of yellow PE
For horizontal storage of steel drums (60 or 200 L capacity)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge