Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Shelving systems for water-endangering and aggressive liquids, KG-GR WPE

Lieferant: LACONT UMWELTTECHNIK

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

R71-2120-D R71-2121-D R71-2122-D R71-2123-D
LACOR71-2120-DEA 0 EUR InStock
LACOR71-2120-D LACOR71-2121-D LACOR71-2122-D LACOR71-2123-D
Shelving systems for water-endangering and aggressive liquids, KG-GR WPE
Möbel Regale
Shelving systems are used for the storage of water-endangering and aggressive liquids.

  • Galvanised shelving parts and tray-shaped shelves with PE insert
  • Height-adjustable shelves at intervals of 25 mm
  • Mixture of basic and extension units allows extension as required
  • Stable construction by reinforced struts

Two different combinable shelf widths (1000 and 1300 mm) allow optimal utilisation of existing storage space.

Maximum unit load: 1200 kg.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge