Jars, wide mouth, conical, graduated
Lieferant: LAMAPLAST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
233 231 238C 238 235C 235 233C 231C
LAMA233EA 3.4 EUR InStock
LAMA233 LAMA231 LAMA238C LAMA238 LAMA235C LAMA235 LAMA233C LAMA231C
Jars, wide mouth, conical, graduated
Dosen Dosen, Universal
PP, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Lid must be ordered separately
Wide mouth universal containers for creams, semi solid substances, crystals, powders and granulates.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge