Jerricans, narrow neck, HDPE, with screwed stopcock
Lieferant: LAMAPLAST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3963 3961S
LAMA3963EA 69.6 EUR InStock
LAMA3963 LAMA3961S
Jerricans, narrow neck, HDPE, with screwed stopcock
Flaschen Kanister
HDPE, natural.
  • Supplied with self sealing plastirob stopcock
  • Self sealing cap
  • Good chemical and temperature resistance
Ideal for use in food industries.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge