Kalibrierthermostate, ECO
Lieferant: LAUDA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
L002847 L002848 L002850
LAUDL002847EA 9780 EUR InStock
LAUDL002847 LAUDL002848 LAUDL002850
Kalibrierthermostate, ECO
Umwälzthermostate Offene Umwälzsysteme Kalibrier-Thermostate
Kalibrier/Bad/Umwälzthermostat mit modernster Microprozessortechnik und integriertem Kühlsystem. Arbeitstemperaturbereich: −25 ... 200 °C.
  • Safety class III for operation with flammable and non-flammable liquids
  • Over-temperature cut-out adjustable via menu
  • Various pump with six adjustable performance levels
  • Easy control of the flow rate between internal and external circulation during operation without contact with the bath
  • USB interface as standard
  • 2-chamber construction for constant level in the thermosetting chamber
  • Vertical adjustment of the overflow height of the thermosetting chamber
  • Bath tank made from stainless steel with drain valve
  • Condenser cooling air
  • Utilises natural refrigerants
The calibration thermostat has a coloured TFT display for simultaneous indication of actual and set values and graphic illustration of the temperature profile. User friendly menu navigation in plain language with an easy input function via cursor and soft keys. Additional Tmax key for overtemperature protection and a fully electronic continuous controller with PID action. USB interface as standard, upgradeable with an interface module (analogue module, contact module, RS232/485 module, Profibus, Ethernet-USB module).
Upgradeable with PT100/LiBus module for external control and remote control via Command console.
Bestellinformation: Calibration racks are available to purchase separately.
Lieferumfang: Supplied with nipples, screw caps and bath cover.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge