Tubings, insulated
Lieferant: LAUDA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LZM052 LZM053 LZM054 LZM055
LAUDLZM052EA 291 EUR InStock
LAUDLZM052 LAUDLZM053 LAUDLZM054 LAUDLZM055
Tubings, insulated
Schläuche Transferschlauch
Metal hoses with insulation for connections with thread M16×1.
  • Temperature resistance: −90 to +150 °C.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge