Cooling water tubing
Lieferant: LAUDA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
RKJ033 RKJ031 RKJ103 RKJ104 RKJ105 RKJ032
LAUDRKJ033EA 44.1 EUR InStock
LAUDRKJ033 LAUDRKJ031 LAUDRKJ103 LAUDRKJ104 LAUDRKJ105 LAUDRKJ032
Cooling water tubing
Schläuche Transferschlauch
EPDM tubing, reinforced.
Special polymer tubings for high pressure. Not suitable for Ultra 350 and mineral oils.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge