Laboratory fume hoods, Protector®, XStream®
Lieferant: LABCONCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
110410021
LBCP110410021EA 0 EUR InStock
LBCP110410021
Laboratory fume hoods, Protector®, XStream®
Dunstabzugshauben und Verkleidungen Chemikalien-Abzüge
The Protector® XStream® benchtop laboratory hoods are chemical resistant, fiberglass reinforced, composite liner and baffle. These are designed for general chemical applications.
  • Durable glacier white, dry powder-coated steel exterior with by-pass flow design
  • One-piece molded fiberglass interior and pre-set baffle with flame spread index less than 25 per ASTM E-84
  • Baffles are removable for cleaning
  • Removable front and side panels and front and interior access panels for access to plumbing and electrical wiring
  • Two tempered safety glass vertical-rising sash with powder-coated aluminum handles and powder-coated steel sash tracks with Clean-Sweep™ slots
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge