Biological safety cabinet, class II, type A2, Purifier® Logic®+
Lieferant: LABCONCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
302481030 302481150 302611040 302681150 302411030 302581041
LBCP302481030EA 0 EUR InStock
LBCP302481030 LBCP302481150 LBCP302611040 LBCP302681150 LBCP302411030 LBCP302581041
Biological safety cabinet, class II, type A2, Purifier® Logic®+
Dunstabzugshauben und Verkleidungen Biologische Sicherheitswerkbänke
Purifier Logic+ Class II, Type A2 Biosafety Cabinets provide personnel, product and environmental protection from hazardous particulates such as agents that require Biosafety Level 1, 2 or 3 containment. Other appropriate applications include work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. These cabinets are designed to work with antineoplastic drugs, genetic material, asbestos and additional substances that generate hazardous airborne particulates. When canopy connected, the biosafety cabinets may be used for applications involving minute quantities of volatile toxic chemicals and tracer amounts of radionuclides as an adjunct to microbiological research.
  • Constant Airflow Profile™ (CAP) Technology
  • Allows the user to program start up and shut down operations
  • Interior mounted, line -of-sight colour LCD information center with easy to understand MyLogic™ OS
  • Ideal for work with hazardous airborne substances
  • Smart-Start™ System
  • Leak-tight stainless steel interior
During operation, room air is drawn into the inlet grill. The balance of the contaminated air is discharged to the environment after passing through the exhaust HEPA filter.
Zertifizierungen: Conforms to AS2252.2, AS1807.22, AS1807.26, NSF listed, CE conformity marking, class 5 conditions per ISO 14644-1 and 2.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge