XPert® TXE™ Equipment Enclosures
Lieferant: LABCONCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3892020 3892021
LBCP3892020EA 11060 EUR InStock
LBCP3892020 LBCP3892021
XPert® TXE™ Equipment Enclosures
Dunstabzugshauben und Verkleidungen Waagen-Abzüge
XPert TXE Equipment Enclosures are designed to address the needs of pharmaceutical research and quality control laboratories.
These vented equipment enclosures may be ducted to the outside or connected to the filtermate portable exhauster for HEPA and/or carbon filtered exhaust back into the laboratory. Specialized base stands are also available (sold separately).
It provides a safe, ventilated space for keeping powder-generating apparatus contained. Its tall 1066,8 mm interior height accommodates equipment such as pill presses, grinders, crushers and sieves. HEPA-filtered air is returned to the laboratory.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge