Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Ductless fume hoods, Paramount®

Lieferant: LABCONCO

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

6963320 6963521 6963623
135-2023EA 0 EUR InStock
135-2023 LBCP6963521 LBCP6963623
Ductless fume hoods, Paramount®
Dunstabzugshauben und Verkleidungen Filterabzüge ohne Abluftleitung
Self-contained ductless enclosures allow safe handling of organic, formaldehyde, acid gas, sulfur compounds, or ammonia chemical fumes and vapors, as well as particulates. Air inside the enclosures passes through carbon filters, and in some cases additional HEPA particulate filters, before being safely exhausted back into the laboratory. No outside ducting is required.

  • Telescoping base stands adjust in seven height positions from 69,9 - 75,1 cm
  • Glacier white stands are constructed of 4,4 cm tubular epoxy-coated steel
  • Hydraulic lift base stands are adjustable to any height from 64,8 - 85,1 cm and include four powder-coated, die-cast feet with levelers
  • Feet can be replaced with casters for mobility
  • Electric hydraulic lift models include a front-accessible up/down switch and a 3,7 m three-wire cord and plug
  • Manual hydraulic lift models include front-accessible hand crank

Containment-enhancing components ensure that no vapors enter the breathing zone, providing a safe work environment. Six different stackable carbon filter types (not included) remove vapor contaminants from the air and can be combined to handle various chemicals and particulates during the same application. Placing a HEPA filter after a carbon filter provides particle-free exhaust suitable for cleanroom applications. An organic vapor sensor detects chemicals breaking through the filters, ensuring operator protection. Digital display shows airflow and filter status and touchpad controls enclosure operation.

Telescoping base stands adjust in seven height positions from 69,9 - 75,1 cm. Glacier white stands are constructed of 4,4 cm tubular epoxy-coated steel. Hydraulic lift base stands are adjustable to any height from 64,8 - 85,1 cm and include four powder-coated, die-cast feet with levelers. Feet can be replaced with casters for mobility. Electric hydraulic lift models include a front-accessible up/down switch and a 3,7 m three-wire cord and plug. Manual hydraulic lift models include front-accessible hand crank.

SoLo™ base stands with remote control are ADA- and NSF-compliant and support a maximum of 589,7 kg. Stands have a depth of 87 cm. Built-in hydraulic lift is operated by remote control providing work surface heights from 43,8 - 93,3 cm and overall enclosure height as low as 193,5 cm. The supported enclosure can be positioned for persons of different statures, wheelchair-bound users, transport through doorways, and easier access to upper components. Stands include 12,7 cm diameter, toe-locking polyurethane casters with bearings for quiet operation. Black, chemical-resistant epoxy work surfaces are dished to contain spills and contoured to fit enclosures. Stainless steel work surfaces feature a 2,5 cm deep front spill trough

Zertifizierungen: Filters must be selected according to individual application requirements and can be stacked two deep. Only two types of filters can be used together, or both can be of the same type. Enclosure meets SEFA 1-2005, ASHRAE 110-95, and ANSI z9.5-2003 standards. CAN/CSA C22.2 No. 1010.1 compliant. UL 3101-1/61010-1 compliant. Also CE marked.

Bestellinformation: Base stand, work surface and filters must be ordered separately.

Order Now
 

PRODUKT

TECHNISCHE DATEN

ZUBEHÖR

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge