Dusting mop, Clean and Away
Lieferant: LEIFHEIT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
56640
LEIF56640EA 0 EUR InStock
LEIF56640
Dusting mop, Clean and Away
Mopps
Floor wiper with screw-on-handle for the dry cleaning of all types of flooring.
  • Comes with telescopic handle (80 - 140 cm) and flexible joint
  • Made for the quick dry cleaning in between
  • Top of the based has four cloth clips to clip the magneto static dust cloth with a simple touch
  • Fexible joint makes it flexible to reach low areas
Recommended to use with Leifheit magneto static cloth.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge