Sleeves for ground joint cones
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01101714 01101719 01101724 01101729 01101745
LENZ01101714EA 25.6 EUR InStock
LENZ01101714 LENZ01101719 LENZ01101724 LENZ01101729 LENZ01101745
Sleeves for ground joint cones
Normschliff-Manschetten und Faltenbälge Schliffmanschetten
Sleeves, made of PTFE. For fitting onto standard ground cones.
  • Very good chemical resistance
  • Gas, liquid and vacuum-tight
  • Temperature resistant from −200 to +280 °C
Sturdy construction with protruding grip so sleeves can be detached easily. With sealing rings on the outside of the ground socket, which prevent ground joints from seizing and give a very good seal. Grease-free connection; no grease residue to contaminate sample.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge