Federn für Aluminiumringe
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01102825 01102835 01102840 01102860
LENZ01102825EA 2.6 EUR InStock
LENZ01102825 801-1731 LENZ01102840 LENZ01102860
Federn für Aluminiumringe
Federn
Stainless steel springs with two rings for the safe connection of conical ground joints in combination with alu rings with hooks.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge