Ball / flange connections, DURAN®
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01250015 01250025 01260015 01260025
LENZ01250015EA 30.7 EUR InStock
LENZ01250015 LENZ01250025 LENZ01260015 LENZ01260025
Ball / flange connections, DURAN®
Adapter Übergangsstücke mit Kugelflanschen
Made from DURAN® tubing.
  • High temperature resistance
  • Very good chemical resistance
Manufactured to inch dimensions in accordance with DIN 12244 and DIN 12264.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge