Screw caps for screw thread tubes
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
01320018 01320014 01320025 01320032 01320045
LENZ01320018EA 3.5 EUR InStock
LENZ01320018 LENZ01320014 LENZ01320025 LENZ01320032 LENZ01320045
Screw caps for screw thread tubes
Verschlüsse Verschlüsse für Reagenz- und Probenröhrchen
PBT,red, with PTFE coated silicone sealing (VMQ).
  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge