Stopcocks, needle-valve, 90° angle
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
02270202
LENZ02270202EA 33.1 EUR InStock
LENZ02270202
Stopcocks, needle-valve, 90° angle
Hähne
Needle-valve stopcocks are made of DURAN® borosilicate glass, with PTFE spindle.
  • Screw cap and retention device made of PP
  • Temperature resistance from −20 to +150 °C
These stopcocks are ideal for exact dispensing and therefore ideal for glass equipment.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge