Vacuum stopcocks
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
02901804 02902106 02962910 02902910 02922910 02901402 02921402 02961402 02961804 02921804 02941402 02941804 02942910 02962106 02922408 02942408 02962408 02902408 02922106 02942106
189-2102EA 51.5 EUR InStock
189-2102 189-2103 LENZ02962910 189-2105 189-2125 LENZ02901402 LENZ02921402 LENZ02961402 LENZ02961804 189-2122 189-2111 189-2112 189-2115 LENZ02962106 189-2124 189-2114 LENZ02962408 LENZ02902408 189-2123 189-2113
Vacuum stopcocks
Hähne
Vacuum stopcocks, DURAN® with side arms and hollow NS glass plug.
  • Excellent chemical resistance
  • Excellent temperature resistance
Leakage rate according to DIN 12540-1.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge