Extractors heads for specific heavy solvents
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
05433158 05433137 05433149
533-2152EA 330 EUR InStock
533-2152 LENZ05433137 LENZ05433149
Extractors heads for specific heavy solvents
Extraktoren Flüssig-Flüssig-Extraktoren
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific heavy solvents, with insert filter (porosity: 0).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge