O-Ring seals for flat flanges
Lieferant: Lenz Laborglas GmbH & CO.KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
06248010 06248015 06248215 06248210 06248206 06248006 06248020 06248220
201-0152EA 15.2 EUR InStock
201-0152 LENZ06248015 LENZ06248215 LENZ06248210 LENZ06248206 201-0151 LENZ06248020 LENZ06248220
O-Ring seals for flat flanges
Versiegeln Dichtringe
Silicone or Silicone PTFE-coated.
  • Excellent chemical resistance
  • Excellent temperature resistance
Seals for flat flanges LF 60-200.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge