Freestanding freezer, with NoFrost, FNd 7227
Lieferant: Liebherr
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FND7227
LIEBFND7227EA 0 EUR InStock
LIEBFND7227
Freestanding freezer, with NoFrost, FNd 7227
Gefriergeräte Gefriergeräte für das Labor und allgemeine Anwendungen
NoFrost protects the freezer compartment from unwanted icing, which consumes a lot of energy and can be expensive. NoFrost means no more tedious and time-consuming defrosting of the freezer compartment, more time for other things and saving money.
  • Frost protect
  • Drawer with comfortable tilting
  • Energy saver
  • Power failure alarm
  • Door alarm
  • Vario space
  • Closing assistance
  • Adjustable feet
  • Replaceable door seal
  • Dishwasher-safe features
The touch display enables easy and intuitive operation. Simple, classic and high-quality with its shiny white-painted surface. You can open the appliance very easily using the vertically integrated and ergonomically fitted handle. Our BluPerformance technology makes it a real all-rounder.
It offers a huge volume and lots of cooling capacity - but also consumes a very small amount of energy. It has such a calm character that you can barely hear it. Its secret lies in its modern interior. All components are technically and mechanically adapted to one another.
Connector cable (length): 2,100 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge