Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

FNf 4605 Pure Freestanding Freezer, with NoFrost

Lieferant: Liebherr

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

FNF4605
LIEBFNF4605EA 0 EUR InStock
LIEBFNF4605
FNf 4605 Pure Freestanding Freezer, with NoFrost
Gefriergeräte Gefriergeräte für das Labor und allgemeine Anwendungen
Simple, classic, high-quality, with its shiny white-painted surface, the Liebherr fits particularly well into its surroundings and transcends fashion and trends, remaining timeless, beautiful and functional. The white surface is robust and easy to clean.

  • Touch display
  • NoFrost
  • Recessed grip
  • Door alarm: Alerts with a pulsating light and a warning tone (door is left open for too long)
  • Adjustable feet
  • Replaceable door seals
  • Handle: Recessed grip (both sides, vertical, continuous)

Refrigerators fit perfectly into a 60 cm deep niche, leaving only the door protruding in a defined manner, ensuring optimum access to the recessed grip and lever handle.

The Bluperformance technology makes it a real all-rounder. It offers a huge volume and lots of cooling capacity and consumes a very small amount of energy.

Ideally set up for such a cool environment because all components are designed to operate in ambient temperatures as low as −15 °C.

Temperature range freezer compartment: −15 to −28 °C
Door opening angle: 115°
Number of temperature zones: 1
Number of regulated refrigeration circuits: 1
Temperature display: Freezer

Door alarm, freezing and SuperFrost are adjustable on appliance and via app. But bottle timer is adjustable on app.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge