Low Temperature Laboratory Chest Freezer, LGT 2325
Lieferant: Liebherr
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LGT2325-43001
LIEBLGT2325-43001EA 0 EUR InStock
LIEBLGT2325-43001
Low Temperature Laboratory Chest Freezer, LGT 2325
Gefriergeräte Ultratiefkühlgeräte
LGT 2325, Low temperature laboratory chest freezer is a top-of-the-line product designed to meet the needs of laboratories, research facilities, and hospitals.
  • Built in lockable door
  • Digital temperature display
  • Static cooling system
  • Climate rating 4 (10 to 30 °C)
  • Energy consumption in 365 days 824 kWH
Bestellinformation: Five years warranty.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge