Performance Black Reach-In Refrigerator with Bottom Compressor
Lieferant: Liebherr
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
MRFVD4011
LIEBMRFVD4011EA 0 EUR InStock
LIEBMRFVD4011
Performance Black Reach-In Refrigerator with Bottom Compressor
Kühlschränke
Digital temperature display: Maintenance free and accurate - easily take accurate temperature readings of the set temperature using the digital temperature display.
  • Replaceable seal
  • Safety glass
  • Forced air cooling
  • Self-closing door
  • Reversible door hinges(left/right)
Housing: Individual and robust: Design the housing exactly as you want it. It is robust, shockproof and easy to clean.
Interchangeable door hinge: The interchangeable door hinge allows you fill or empty your appliance in any place.
Recessed grip: Easy handling - you can open and close the door easily using the recessed grip fitted on the side of the door. Nothing sticks out, so there's no chance of snagging yourself on anything. A huge advantage, even in narrow spaces.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge