Slide box
Lieferant: LIEDER JOHANNES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PK25
LIEDPK25EA 5.7 EUR InStock
LIEDPK25
Slide box
Objektträger-Kästen
This slide box is designed to facilitate the transportation of 25 microscope slides.
  • Transparent cover for easy viewing
  • Robust design and stackable construction
  • Serrated retainer strips to maintain slides properly
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge