TBX agar
Lieferant: LIOFILCHEM
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10522
LIOF10522EA 34.3 EUR InStock
LIOF10522
TBX agar
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
X-glucuronide, contained in the TBX agar formulation, is the chromogenic agent that allows the determination of the β-Dglucuronidase activity, which is a highly specific enzyme for E. coli. Gram-positive bacteria are inhibited by bile salts.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge