Photometer, MD 600 with reagents for waste water
Lieferant: TINTOMETER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
214100
LOVI214100EA 3091.1 EUR InStock
LOVI214100
Photometer, MD 600 with reagents for waste water
Umwelt-Prüfkits
MD 600 photometer is for precise and accurate measurements with a small footprint.
  • Complete and ready to use system
  • Ideal for site laboratories where accuracy, simplicity and space are key requirements
The setup uses Lovibond ® Vario tube tests and powder packs which enable the user to easily perform highly sensitive and precise water testing. It tests COD LR(0 to 150 mg/L), COD MR(0 to 1500 mg/L), Ammonia HR(1 to 50 mg/L N), Nitrate (1 to 30 mg/L), Nitrite (0,01 to 0,3 mg/L N), Nitrogen (Total 5 to 150 mg/L N) and Phosphate (0,02 to 1 mg/L P).
Lieferumfang: It is supplied with MD 600 photometer and its accessories, RD 125 thermoreactor, IRiM infra-red module, tube, vial stand, membrane filter set, instruction manual and guarantee sheet.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge