Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Drum pumps sets for hazardous fluids

Lieferant: LUTZ PUMPEN

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

0205-411
LUTZ0205-411EA 4160 EUR InStock
LUTZ0205-411
Drum pumps sets for hazardous fluids
Pumpen Fass/Transferpumpen
This pump set is specially adapted for pumping hazardous fluids or solvents such as acetone, formic acid, butyl acetate, acetic acid, nicotine, methyl benzene (toluol) and styrol.

  • Fast assembly
  • Immediately ready for action
  • Ideal for pumping and filling thin bodied liquids
  • Compact compressed air motor powerful and reliable

The explosion proof universal motor ME II 3 is ideally suitable for transferring of many thin bodied easily flammable and combustible liquids.

Zertifizierungen: Explosion proof according to ATEX directive 94/9/EC category 2.

Lieferumfang: Pump set includes 1 Motor ME II 3, 1 pump tube SS 41-L-SL stainless steel, 2,5 m solvent hose 1", electrically conductive bound with hose connectors brass, nozzle brass, equipotential bonding cable and drum adapter PP.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge