Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid 98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

M2128-5G M2128-100MG M2128-10G M2128-1G M2128-250MG M2128-500MG
M2128-5GEA 442 EUR InStock
M2128-5G M2128-100MG M2128-10G M2128-1G M2128-250MG M2128-500MG
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid 98%, Sigma-Aldrich®
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
Thiazolyl Blue Tetrazolium Blue (MTT) may be used in measurement of cell proliferation. MTT produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells. The blue crystals are solubilized with acidified isopropanol and the intensity is measured colorimetrically at 570 nm. MTT has been used as a histochemical/cytochemical reagent and for the detection of NAD. ADP-linked enzyme systems in tissue cannot be detected with MTT, due to binding of the cation by the cyanide trap used. MTT is rapidly reduced to the formazan, which chelates with nickel, copper, and cobalt; the cobalt chelate has been used in oxidative systems.

  • Thiazolyl Blue Tetrazolium Bromide has been used for the following assays:
  • Proliferation assay
  • TNF (tumor necrosis factor) bioassay


Formel: C₁₈H₁₆N₅BrS
MW: 414.33 g/mol
Melting Pt: 195 °C (Zers., Lit.)
Storage Temperature: Kühlschrank
MDL Number: MFCD00011964
CAS-Nummer: 298-93-1
EINECS: 206-069-5
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561211eb7a01c80370a0145ebbb045631152528d&vid=561211eb7a01c80370a0145ebbb045631152528d&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Purity (HPLC) ≥98.0%
Solubility (Turbidity), c = 5 mg/ml (0.5%); Water Clear
Solubility (Colour) Yellow to Very Dark Yellow and Yellow-Green
Recommended Retest Period 5 years

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge