VWR® Protease Inhibitor Cocktail, Plant
Lieferant: VWR Chemicals
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
M307-1MLEA 119 EUR InStock
M307-1ML
VWR® Protease Inhibitor Cocktail, Plant
Zellkultur-Ergänzungen und -Zusätze
Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, Pepstatin, Leupeptin and 1, 10-Phenanthroline. Each vial can be reconstituted in 1 ml dimethyl sulfoxide to form a 100X solution.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge