Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

4,4'-Bis(dimethylamino)diphenylmethan 98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ100

M44451-100G M44451-500G M44451-5G
M44451-100GEA 177 EUR InStock
M44451-100G M44451-500G M44451-5G
4,4'-Bis(dimethylamino)diphenylmethan 98%, Sigma-Aldrich®
4,4'-Bis(dimethylamino)diphenylmethan
4,4′-Methylenebis(N,N-dimethylaniline) is an intermediate in dye manufacture and works as a reagent for the determination of lead. It is suitable for the qualitative assay of cyanogens.

  • Application
  • 4,4′-Methylenebis(N,N-dimethylaniline) has been used to check the production of hydrogen cyanide by bacteria
  • 4,4′-Methylenebis(N,N-dimethylaniline) has been used as a component in the preparation of Feigl-Anger paper for visualising HCN release
  • It may also be suitable for the preparation and characterisation of inclusion complex with β-cyclodextrin

4,4′-Methylenebis(N,N-dimethylaniline) is used in the adduct purification of precursors. It is used as an intermediate in the preparation of its hydrochloric salt.


Formel: C₁₇H₂₂N₂
Melting Pt: 88,..89 °C (Lit.)
Flash-Pt: 178 °C (geschlossener Tiegel)
Storage Temperature: Room Temperature
MDL Number: MFCD00008317
CAS-Nummer: 101-61-1
EINECS: 202-959-2
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7bea99de38721d9fe8fcdcb8e6d8601df58bcb4&vid=d7bea99de38721d9fe8fcdcb8e6d8601df58bcb4&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour); White to Faint Beige to Tan Conforms to Requirements
Appearance (Form); Powder, Flakes, Crystals, Crystalline Powder and/or Chunks Conforms to Requirements
Infrared spectrum Conforms to Structure
Purity (Titration by HClO₄) 97.5 - 102.5 %
Purity (GC) ≥97.5%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge