Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Methylgrün-Zinkchlorid-Doppelsalz ~85%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Gefahr

M-Clarity™ Qualitätsstufe = MQ200

M8884-25G M8884-5G
M8884-25GEA 266 EUR InStock
M8884-25G M8884-5G
Methylgrün-Zinkchlorid-Doppelsalz ~85%, Sigma-Aldrich®
Methylgrün-Zinkchlorid-Doppelsalz
Methyl green is a synthetic, triphenylmethane-type cationic dye related to ethyl green mainly used as DNA stain in histochemistry. It possesses aniline rings with different degrees of methylation. The dye binds specifically to the A-T rich regions of the major groove of DNA with the help of two positive charges. Methyl green in combination with pyronin is extensively used to stain RNA and plasma cells in tissue sections and cytologic preparations.

Methyl green is suitable as a nuclear counterstain in enzyme histochemistry, immunostaining, and in situ hybridisation as the bluish-green color will be in contrast with red, purple, brown, and black products of histochemical reactions. It is also suitable as a tracking dye in Acid-polyacrylamide gel electrophoresis to separate gliadins. It has been used in the histochemical staining for the apoptosis examination of kidney tissue and as a nuclear stain for labeling embryos. Material may be a mixture of monobromo-monochloro and dichloro ZnCl₂ salts. Halogen contents listed on label.


Formel: C₂₇H₃₅BrClN₃·xZnCl₂
Melting Pt: >300 °C (Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00151094
CAS-Nummer: 7114-03-6
EINECS: 230-415-4
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=914ee3058a766a160715eabb0f667912997c3ed2&vid=914ee3058a766a160715eabb0f667912997c3ed2&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Red to Very Dark Red and Red-Brown and Brown-Red
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wave Length, 0,005 G/L H₂O 626 - 634 nm
Extinction Coefficient ≥16000
Wave Length 311 - 319 nm
Solubility (Colour), C = 1 mg/ml, H₂O Dark Blue to Very Dark Blue and Dark Blue-Green to Very Dark Blue-Green
Dye Content ≥83 %

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge