Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

L(+)-Arginin 98,5-101,0% USP, Multikompendial, erfüllt die analytischen Spezifikationen von ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™

Lieferant: MACRON AVANTOR BRAND

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


MACR4926-06EA 0 EUR InStock
MACR4926-06 MACR4926-07 MACR4926-09
L(+)-Arginin 98,5-101,0% USP, Multikompendial, erfüllt die analytischen Spezifikationen von ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
L(+)-Arginin


Formel: C₆H₁₄N₄O₂
MW: 174.2 g/mol
Siedepunkt: 368 °C
Melting Pt: 238 °C
Dichte: 1,3 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00002635
CAS-Nummer: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785
Order Now
Spezifikation Testergebnisse
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets J.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
Preserve in Tight Containers
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (C6Hl4N402) (dried basis) 98.5 - 101.5%
USP - Identification Passes Test
USP - Specific Rotation (A]25 AO (+) 26.3 - 27.7°
USP - Loss on Orying at 105 °C ≤0.5%
USP - Chloride (Cl) ≤0.05%
USP - Residue on Ignition ≤0.3%
USP - Sulfate (S04) ≤0.03%
USP - Iron (Fe) ≤0.003%
USP - Related Compounds - Individual Impurities ≤0.5%
USP - Related Compounds - Total Impurities ≤2.0%
EP/BP - Assay (C6Hl4N402) (dried basis) 98.5 - 101.0%
EP/BP - Identification A Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Specific Rotation (A]2°AO (+) 25.5 - 28.5 °
EP/BP-Ninhydrin-Positive Substances-Each ≤0.2%
EP/BP-Ninhydrin-Positive Substances-Total Impurities ≤0.5%
EP/BP - Chloride (Cl) ≤200ppm
EP/BP - Sulfate (S04) ≤300 ppm
EP/BP - Ammonium (NH4) ≤0.02%
EP/BP - Iron (Fe) ≤10 ppm
EP/BP - Loss on Orying ≤0.5%
EP/BP - Ash (sulfated) ≤0.1%
JP - Identification Passes Test
JP - 0ptical rotation, 20 °C (+) 26.9 - 27.9°
JP - pH (1:10) 10.5 - 12.0
JP - Clarity and Color of Solution Passes Test
JP - Chloride (Cl) ≤0.021%
JP - Sulfate (S04) ≤0.028%
JP - Ammonium (NH4) ≤0.02%
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Iron (Fe) ≤10 ppm
JP - Related Substances Passes Test
JP - Loss on Orying at 105 °C ≤0.30%
JP - Residue after Ignition ≤0.1%
JP - Assay (C6Hl4N402)(dried basis) 98.5 - 101.0%
Endotoxin Concentration, IU/mg, For Information only
ChP - Specific Rotation @ 20 °C 26.9 - 27.9
ChP - Identification 1 (TLC) Passes Test
ChP - Identification 2 (IR) Passes Test
ChP - Alkalinity (pH) 10.5 - 12.0
ChP - Transmittance of solution 0.98
ChP - Chlorides ≤0.02%
ChP - Sulfates ≤0.02%
ChP - Ammonium ≤0.02%
ChP - Proteins Passes Test
ChP - 0ther amino acids (TLC)(<=0.4%, one spot only) Passes Test
ChP - Loss on drying @ 105 °C ≤0.5%
ChP - Residue on ignition ≤0.1%
ChP - Iron ≤0.001%
ChP - Heavy metals ≤0.001%
ChP - Arsenic ≤0.0001%
ChP - Assay, dried basis 99.0 - 101.0%

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge