Phase separating paper
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
484011 484015 584011 584015 584012 430009 430011 430012 430015 430018 430024 484005 484007 484009 484012 484018 484024 484032 584009 584018 584024 584032
MANA484011EA 23 EUR InStock
MANA484011 MANA484015 MANA584011 516-6012 516-6019 MANA430009 MANA430011 MANA430012 MANA430015 MANA430018 MANA430024 MANA484005 MANA484007 MANA484009 MANA484012 MANA484018 MANA484024 MANA484032 MANA584009 MANA584018 MANA584024 MANA584032
Phase separating paper
Filter Filterpapiere Phasentrennpapiere
These papers are made hydrophobic (impermeable to water) by impregnation with a silicone. With the aid of these filters, water can be separated from water-immiscible organic solvents in an elegant manner, by means of a simple filtration.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge