Filter papers for fat analysis
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
528005 528007 528009 528011 528012 528015 528018 528024 528027 561032 591005 591007 591009 591011 591012 591015 591018 591024 591027
MANA528005EA 31 EUR InStock
MANA528005 MANA528007 MANA528009 MANA528011 MANA528012 MANA528015 MANA528018 MANA528024 MANA528027 MANA561032 MANA591005 MANA591007 MANA591009 MANA591011 MANA591012 MANA591015 MANA591018 MANA591024 MANA591027
Filter papers for fat analysis
Filter Filterpapiere Qualitatives Filterpapier
Smooth folded filter papers for fat analysis.
MN 615 ff paper is particularly suited for the analysis of fats. A special treatment with organic solvents guarantees that these filters are practically free of fats and resins (ether-soluble residue < 0,1 mg for a 270 mm filter circle).
MN 715 paper is also suited for the analysis of fats. Careful selection of the raw materials ensures a low ether-soluble residue for these filters.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge