Qualitative filter papers, grades MN 616 md and 616 md 1/4, circles and folded filters
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
433011 433005 433007 433009 433012 433015 433018 433024 433032 533005 533007 533009 533011 533012 533015 533018 533024 533032
MANA433011EA 14.2 EUR InStock
MANA433011 MANA433005 MANA433007 MANA433009 MANA433012 MANA433015 MANA433018 MANA433024 MANA433032 MANA533005 MANA533007 MANA533009 MANA533011 MANA533012 MANA533015 MANA533018 MANA533024 MANA533032
Qualitative filter papers, grades MN 616 md and 616 md 1/4, circles and folded filters
Filter Filterpapiere Qualitatives Filterpapier
MN 616 md is slightly more retentive than MN 615 with a corresponding increase in filtration time.
  • Slightly more retentive than standard grades
  • For routine analysis
  • Medium flow rate
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge