Shell vials, N 8 and N 12
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
70202.1
MANA70202.1EA 13.9 EUR InStock
MANA70202.1
Shell vials, N 8 and N 12
Vials
These shell vials for HPLC analyses have the advantage of an easy handling.
  • Made of clear and amber glass
  • Flat bottom
  • Fire-polished neck
Useable for the HPLC autosamplers of Shimadzu, VWR® (Merck / Hitachi - only for Chromaster, LaChrom Elite L-2200, L2200U) and Waters (only for WISP, 717(Plus), GPC 2000).
PE plugs with a diaphragm for safe penetration of the needle are used for the shell vials.
Easy and quick to seal the vial/closure combination.
These easy to handle vial/closure combinations are only suitable in cases of low requirements on tightness, as the plugs have no elastomeric sealing element.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge