HPLC columns, NUCLEOSIL® C18
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
720041.40 720051.40 720730.46 720740.46 720949.46 721070.30 721074.20 721074.30 721075.30 720002.20 720002.30 720002.40 720002.46 720014.20 720014.30 720014.40 720014.46 720018.40 720018.46 720023.40 720023.46 720040.40 720040.46 720041.46 720042.40 720043.40 720043.46 720051.46 720055.40 720055.46 720065.40 720065.46 720074.46 720077.46 720098.46 720120.46 720133.40 720133.46 720150.40 720150.46 720120.40 720841.46 720149.46 720141.46 720141.40 720110.46 720951.46 720149.40 721022.30 720140.46 720701.46 721085.30 721473.30
MANA720041.40EA 450 EUR InStock
MANA720041.40 MANA720051.40 MANA720730.46 MANA720740.46 MANA720949.46 MANA721070.30 554-5795 554-5796 MANA721075.30 MANA720002.20 MANA720002.30 MANA720002.40 MANA720002.46 554-5791 554-5792 554-5793 554-5794 MANA720018.40 MANA720018.46 MANA720023.40 MANA720023.46 MANA720040.40 MANA720040.46 MANA720041.46 MANA720042.40 MANA720043.40 MANA720043.46 MANA720051.46 MANA720055.40 MANA720055.46 MANA720065.40 MANA720065.46 MANA720074.46 MANA720077.46 MANA720098.46 554-5790 MANA720133.40 MANA720133.46 MANA720150.40 MANA720150.46 554-5789 MANA720841.46 MANA720149.46 MANA720141.46 MANA720141.40 MANA720110.46 MANA720951.46 MANA720149.40 MANA721022.30 MANA720140.46 MANA720701.46 MANA721085.30 MANA721473.30
HPLC columns, NUCLEOSIL® C18
Chromatographiesäulen
NUCLEOSIL® C18 are octadecyl modified silica phases for HPLC.
  • Robust spherical silica gel
  • Hydrophobic, weakly polar, endcapped phases for RP chromatography
  • Separation mechanism based on hydrophobic (van der Waals) and slight residual silanol interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
Verpackung: Standard NUCLEOSIL® C18 phases are available as packed EC columns for analytical HPLC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge