SPE columns, CHROMABOND® C2
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
730221
MANA730221EA 170 EUR InStock
MANA730221
SPE columns, CHROMABOND® C2
Chromatographiesäulen
CHROMABOND® C2 is a dimethyl modified silica phase for SPE.
  • Very polar RP phase
  • Slight hydrophobic and strong polar interactions
  • Suitable for polar substances with hydrophobic areas from polar matrices
Recommended for biological molecules.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge