Nitrile Palm Coated Thermal Gloves, Temp-Dex 710
Lieferant: MAPA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
34710039
MAPA34710039EA 7.7 EUR InStock
MAPA34710039
Nitrile Palm Coated Thermal Gloves, Temp-Dex 710
Handschuhe
Gloves, Temp-Dex 710, high dexterity and thermal protection, hi-vis yellow seamless knit, black sandy Nitrile with embossed dots, knit wrist, contact heat resistant up to 212 °F.
  • Thin anatomically-shaped glove ensures dexterity
  • Suitable for long-term wear: excellent abrasion resistance
  • Silicone free to prevent traces, defects on metal sheet and glass prior to painting
  • Dot embossing for better insulation and better grip
Zertifizierungen: EN407 X1XXXX, EN388 4111X.
Verpackung: Sizes 7, 9, 11, 10 pairs per bag, 50 pairs per case.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge