Microscope slide, UniMark®
Lieferant: MARIENFELD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0704022
MARI0704022EA 421.5 EUR InStock
MARI0704022
Microscope slide, UniMark®
Objektträger Mikroskop-Objektträger
UniMark slide is designed with a white circle of Ø 12 mm and a white marking area of 20 mm for use in cytocentrifuges.
It is suitable for suitable for marking with inkjet of laser printers and permanent markers.
Zertifizierungen: DIN 8037/1. IVD 98/79/EC.
Verpackung: 50 pieces in a box.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge