Adhesion slide, HistoBond® +S
Lieferant: MARIENFELD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0810501
MARI0810501EA 648.8 EUR InStock
MARI0810501
Adhesion slide, HistoBond® +S
Objektträger Adhäsions-Objektträger
This adhesion slide has been designed for mounting tissue sections.
  • Autoclavable
  • Marking area
  • 90° ground edges
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge