Wax plates for sealing capillary tubes
Lieferant: MARIENFELD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2960409
MARI2960409EA 28.2 EUR InStock
MARI2960409
Wax plates for sealing capillary tubes
Versiegeln Versiegelungsmaterial
Wax plates are used for sealing capillary tubes for micro hematocrits.
  • Two deposit areas on a plastic base
  • Numbered up to 48
Contain modelling clay and additives of food filler material and natural bees wax. We consciously forego adding any chemicals which could affect the plastic material after some time.
Verpackung: Packed in poly bags.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge