Pasteur pipettes, disposable
Lieferant: MARIENFELD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3233050 3233049
MARI3233050EA 71.3 EUR InStock
MARI3233050 MARI3233049
Pasteur pipettes, disposable
Pipetten Pasteur- und Transfer-Pipetten
Pasteur pipettes made of soda-lime glass.
  • For single use
  • Long, fine tip
  • Constricted upper suction end to retain a cotton plug (supplied without cotton plug)
  • Handy packaging in boxes containing 250 pieces
Zertifizierungen: In compliance with DIN ISO 7712
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge