Test tubes with thread
Lieferant: MARIENFELD
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3704679
MARI3704679EA 0 EUR InStock
MARI3704679
Test tubes with thread
Röhrchen Reagenzgläser- und Probenröhrchen
Soda-lime glass.
  • Good chemical and physical properties
  • Without screw cap
Culture medium tubes with thread for aluminium screw caps are suitable for transporting and growing of cultures.
Bestellinformation: Screw caps must be ordered seperately.
Order Now
PRODUKT
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge