Safety scissors, SECUMAX 564
Lieferant: MARTOR ARGENTAX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
564001.00
MARO564001.00EA 58.6 EUR InStock
MARO564001.00
Safety scissors, SECUMAX 564
Scheren
The SECUMAX 564 is characterised by its thumb opening and its extra-long edge, which also makes an ideal support surface. These safety scissors enable you to completely control your work.
  • Sharp, but not dangerous
  • Non-rusting stainless steel
  • Extra-long cutting edges
  • Robust, fiber-glass reinforced plastic handle
Highest safety guaranteed by the special sharpening of the cutting blades and the rounded tips.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge