Petri dish safety dispenser
Lieferant: MBV
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
00.1000.01
MBVA00.1000.01EA 122 EUR InStock
MBVA00.1000.01
Petri dish safety dispenser
Gestelle Spender für Petrischalen
PP dispenser for Petri dishes and RODAC plates.
  • Stackable
  • Absolute safety during transport
  • Autoclavable
The PSD-10 can be used in all microbiological laboratories. It serves to prevent lids accidental falling off and so protects the user and the environment from microorganisms and potentially toxic and dangerous contamination. It offers a safe way to transport Petri dishes and RODAC plates. Each Petri dish is individually packed in a container. The lid of the dish is pressed against the bottom part and thus it is impossible for the lid to fall off. By pressing on the colour clip, the plate can easily be removed by hand.
Dimensions of the container: 103×100×21 mm
Weight: ~38 g
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge