Penicillin/Streptomycin/Amphotericin B -Lösung, Corning®
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30-004-CI
MDTC30-004-CIEA 238.88 EUR InStock
MDTC30-004-CI
Penicillin/Streptomycin/Amphotericin B -Lösung, Corning®
Penicillin/Streptomycin/Amphotericin B
This is a combination of penicillin, streptomycin and Amphotericin formulated to contain 10000 units/ml penicillin G, 10 mg/ml streptomycin sulphate and 25 µg/ml amphotericin B.
Penicillin acts by inhibiting bacterial cell-wall synthesis, Streptomycin inhibits prokaryote protein synthesis, and Amphotericin B interferes with fungal membrane permeability.
This combination is effective against Gram-negative and Gram-positive bacteria, fungi and yeasts. The form provided by Mediatech is solubilised in a proprietary citrate buffer and is recommended for use in cell culture applications at 10 ml/L.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge