Foetal bovine serum, South American origin, Corning®
Lieferant: Corning Life Science
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
35-079-CV
MDTC35-079-CVEA 410.27 EUR InStock
MDTC35-079-CV
Foetal bovine serum, South American origin, Corning®
Seren
Foetal bovine serum sourced in South America and manufactured at facilities approved by the appropriate local authorities.
  • Triple sterile filtered (0,1 μm)
  • Low endotoxin (<30 EU/ml)
  • Low haemoglobin (<30 mg/dl)
Corning® sera products are analysed for quality and performance. Testing includes pH, osmolality, growth promotion, sterility (current USP method), mycoplasma, total protein, endotoxin and haemogloblin levels, and viral contamination. A full biochemical profile is available for each lot upon request.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge