Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, without phenol red, Corning®
Lieferant: MEDIATECH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
40-102-CV
MDTC40-102-CVEA 205.42 EUR InStock
MDTC40-102-CV
Serum-free/low-protein medium, 1X with L-glutamine and 1 g/L BSA, without phenol red, Corning®
Zellkulturmedien
Corning® cellgro® COMPLETE formulation is based on a combination of 1:1 DMEM/F12, RPMI, and McCoy's 5A medium. This medium contains a proprietary mixture of selected trace elements and high molecular weight carbohydrates, extra vitamins, animal origin-free protein, and a small amount of high quality bovine serum albumin (1,0g/L). The Corning cellgro COMPLETE supports superior growth and viability over long-term passages in hybridomas, along with suspensions and adherent cell cultures.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge